Learn More
Trajan Scientific Cleaning Kit Needle Kit Medical 1/EA
SDP
Catalog No. 5000493883
Encompass_Preferred
Needle cleaning kit (Stylet wires 15 cm x 0 10 mm OD PK5 15 cm x 0 15 mm OD PK5 15 cm x 0 20 mm OD PK5 20 mL cleaning solution concentrate tweezers)
Catalog No. 50-004-93883 Supplier Trajan Scientific Supplier No. 031782
Needle cleaning kit (Stylet wires 15 cm x 0 10 mm OD PK5 15 cm x 0 15 mm OD PK5 15 cm x 0 20 mm OD PK5 20 mL cleaning solution concentrate tweezers)