Learn More
SKYLINE CENTER Item #W11 Bulk 2 1/2 Square -1000 ct.
Catalog No. NC1448924
Encompass
2 1/2" SQUARE 100% COTTON PATCHES Uses: Cleaning - guns, firearms, melt flow index, melt plastometers, capillary rheometers, optics, laboratory equipment. Gun Cleaning - 20 - 12 Gauge
This item is not returnable. View return policy
Catalog No. NC1448924 Supplier SKYLINE CENTER Supplier No. W11 1000
2 1/2" SQUARE 100% COTTON PATCHES Uses: Cleaning - guns, firearms, melt flow index, melt plastometers, capillary rheometers, optics, laboratory equipment. Gun Cleaning - 20 - 12 Gauge