Learn More
Roche Diagnostics Cleaning Solution Refill 4 X60 ml
Catalog No. 50589353 Shop All Roche Diagnostics Products
Encompass
Cedex Cleaning Solution is required for cleaning of the capillary system of the Cedex and Cedex HiRes Systems. The ready-to-use solution is certified and available as a component of the Cedex Reagent Kit and Cedex HiRes Reagent Kit and as a separate bottle. For use in quality control/manufacturing process only. 4 x 60ml
Catalog No. 50-589-353 Supplier Roche Diagnostics Supplier No. 05650666001
Cedex Cleaning Solution is required for cleaning of the capillary system of the Cedex and Cedex HiRes Systems. The ready-to-use solution is certified and available as a component of the Cedex Reagent Kit and Cedex HiRes Reagent Kit and as a separate bottle. For use in quality control/manufacturing process only. 4 x 60ml