Learn More
HANNA INSTRUMENTS INC General Purpose pH Electrode Cleaning Solution (500 mL)
SDP
Catalog No. NC1814910
Encompass
HI7061L is a general purpose cleaning solution for use with pH and ORP electrode that can be used for a variety of applications. pH electrodes become dirty with use and can produce inaccurate results. Use our General Purpose pH Electrode Cleaning Solution to prolong the life of your electrode and ensure accurate readings.Specially formulated for cleaning pH and ORP electrodesClearly marked expiration date and lot number for traceability.Air-tight seal to ensure the quality of the solution
Catalog No. NC1814910 Supplier HANNA INSTRUMENTS INC Supplier No. HI7061L
HI7061L is a general purpose cleaning solution for use with pH and ORP electrode that can be used for a variety of applications. pH electrodes become dirty with use and can produce inaccurate results. Use our General Purpose pH Electrode Cleaning Solution to prolong the life of your electrode and ensure accurate readings.Specially formulated for cleaning pH and ORP electrodesClearly marked expiration date and lot number for traceability.Air-tight seal to ensure the quality of the solution