Learn More
MSC Rite-Kem Cleaning Solution, Liquid Concentrate, Pail, 5 Gallon, Non-Acid
Catalog No. 502836564
Encompass
Rite-Kem THIS-N-THAT Cleaning Solution is a versatile liquid concentrate designed for effective porcelain and general bathroom cleaning. It efficiently removes dirt, body oils, stains, and soap scum from various surfaces, while being safe for bathroom materials. The solution can be utilized at full strength for extreme cleaning tasks or diluted for routine daily maintenance.
  • Liquid concentrate formula
  • Effective for porcelain and general bathroom cleaning
  • Removes dirt, body oils, stains, and soap scum
  • Safe on bathroom surfaces
  • Non-acidic formulation
  • Pleasant scent
  • Packaged in a 5 gallon pail
  • Use full strength for extreme cleaning
  • Dilutable to 1-10 for daily maintenance
Catalog No. 50-283-6564 Supplier MSC Supplier No. 48175533
Rite-Kem THIS-N-THAT Cleaning Solution is a versatile liquid concentrate designed for effective porcelain and general bathroom cleaning. It efficiently removes dirt, body oils, stains, and soap scum from various surfaces, while being safe for bathroom materials. The solution can be utilized at full strength for extreme cleaning tasks or diluted for routine daily maintenance.
  • Liquid concentrate formula
  • Effective for porcelain and general bathroom cleaning
  • Removes dirt, body oils, stains, and soap scum
  • Safe on bathroom surfaces
  • Non-acidic formulation
  • Pleasant scent
  • Packaged in a 5 gallon pail
  • Use full strength for extreme cleaning
  • Dilutable to 1-10 for daily maintenance