Learn More
ELITechGroup Thermocouple Cleaning Solution, 60mL
SDP
Catalog No. NC9600953
Encompass
This item has a minimum order quantity of 3 per supplier requirements. Ammonium Hydroxide, Aqueous Solution; Liquid, Clear to pale yellow, distinct, pungent odour; Cleaning Solution (for cleaning thermocouples); B.P. ~100 deg.C; Freezing Point ~0 deg.C; Storage: RT away from direct light
Catalog No. NC9600953 Supplier ELITechGroup Supplier No. SS003
This item has a minimum order quantity of 3 per supplier requirements. Ammonium Hydroxide, Aqueous Solution; Liquid, Clear to pale yellow, distinct, pungent odour; Cleaning Solution (for cleaning thermocouples); B.P. ~100 deg.C; Freezing Point ~0 deg.C; Storage: RT away from direct light