Learn More
Hach Company Cleaning Solution for PHOSPHAX sc Analyzer, 1000 mL
Catalog No. NC3910047 Shop All Hach Company Products
Encompass
Cleaning solution formulated for use with PHOSPHAX sc analyzers (high and low range). Supplied in a 1000 mL bottle, it removes deposits and residues to help maintain accurate phosphate measurements and reliable instrument performance. Consult the Safety Data Sheet (SDS) and follow the manufacturer's handling instructions prior to use.
  • Designed for periodic maintenance and cleaning of the analyzer's measurement chamber and flow paths.
  • Supplied as a 1000 mL ready-to-use solution.
  • Removes scale, deposits, and sample residues that can affect measurement accuracy.
  • May be subject to hazardous shipping; consult the product Safety Data Sheet (SDS) before use.
  • Compatible with both high-range and low-range analyzer configurations.
Catalog No. NC3910047 Supplier Hach Company Supplier No. 2825352
Cleaning solution formulated for use with PHOSPHAX sc analyzers (high and low range). Supplied in a 1000 mL bottle, it removes deposits and residues to help maintain accurate phosphate measurements and reliable instrument performance. Consult the Safety Data Sheet (SDS) and follow the manufacturer's handling instructions prior to use.
  • Designed for periodic maintenance and cleaning of the analyzer's measurement chamber and flow paths.
  • Supplied as a 1000 mL ready-to-use solution.
  • Removes scale, deposits, and sample residues that can affect measurement accuracy.
  • May be subject to hazardous shipping; consult the product Safety Data Sheet (SDS) before use.
  • Compatible with both high-range and low-range analyzer configurations.