Learn More
IMEB, Inc Pathology Bone Band Saw Optional Cleaning Store for Diamond Blades
SDP
Catalog No. NC0907427
Encompass_Preferred
Cleaning Stone for Diamond Blades for the IMEB Bone Band Saw. Bone Band Saw sold separately. The bone band saw is an efficient and economical way of cutting bone in laboratory. The bone band saw only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.
Catalog No. NC0907427 Supplier IMEB, Inc Supplier No. CS90
Cleaning Stone for Diamond Blades for the IMEB Bone Band Saw. Bone Band Saw sold separately. The bone band saw is an efficient and economical way of cutting bone in laboratory. The bone band saw only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.