Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cleaning Up Oil Spills Naturally
SCI_ED

Catalog No. S23196
Change view
Click to view available options
Product Type:
Cleaning Up Oil Spills
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S23196 Cleaning Up Oil Spills
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S23196 Supplier Neo/Sci Corporation Supplier No. 201253

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Ready-to-use bacterial mixture, oil, Petri dishes, vials, pipets, measuring spoon, stirring sticks, sand, gravel, cobbles, gloves, magnifiers and comprehensive teacher and student guides

Requires

Compound microscope

Simulate an oil spill in water and treat it with a microbial mix to observe the physical changes and degradation of oil film.

  • Three spill scenarios: beach sand, gravel and cobbles
  • Observe differences in degradation rates
  • Explore bioremediation methods for other waste chemicals
  • Compare their effectiveness to traditional cleanup methods

Order Info

Class size: 40 students.

Specifications

For Use With (Application) To observe physical changes and degradation of oil film
Includes Ready to use bacterial mixture, oil, petri dishes, vials, pipets, measuring spoon, stirring sticks, sand, gravel, cobbles, gloves, magnifiers and comprehensive teacher and student guides
Class Size 40 Students
Product Type Cleaning Up Oil Spills
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.