Learn More
Grainger Grainger
Supplier:  Grainger 45TW63
Encompass_Preferred
The Grainger 4LUM6 LED utility light is an industrial grade portable work light that provides 360 degrees of bright, energy efficient illumination. It features 108 high-power LEDs with a light output of 6000 lumens and a color temperature of 5000K daylight white. The light is impact resistant, operates on 120V AC power or a rechargeable battery, and has a lightweight polymer housing with an adjustable metal stand that allows it to be hung or positioned as needed. With an IP65 rating, the utility light is dust tight and protected against water jets from any direction.
Catalog No. NC2207602