Learn More
LGI CLEANROM WH 3.37X2/15 PVC CARD
Catalog No. 501126978
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
White cleanroom card from Lgi. ISO 9001-2000 certified. RoHS compliant. 1k/pk. Min. order: 4 pks. Lgi also creates innovative custom labels that meet your specific cleanroom requirements. Based on their proprietary CleanMark process, engineers & designers will work w/ you to design & construct labels to fit any size, permanent or removable adhesive, direct thermal or thermal transfer printable, can withstand extreme temperatures, harsh chemicals & abrasion. Labels can indicate sterilization such as autoclave or gamma or can be pre-sterilized. 100% synthetic, non-particulating, ultra low-outgassing raw materials wont contaminate clean or sterile environments. Lgis exclusive EZ-Peel feature is adhesive-free on one or both sides of the label, making it easy to apply & remove w/ protective gloves typical in cleanrooms, fabs, medical labs & other controlled environments.
Catalog No. 50-112-6978 Supplier LGI Supplier No. 10291V
This item has a minimum qty of 3 per supplier requirements.
White cleanroom card from Lgi. ISO 9001-2000 certified. RoHS compliant. 1k/pk. Min. order: 4 pks. Lgi also creates innovative custom labels that meet your specific cleanroom requirements. Based on their proprietary CleanMark process, engineers & designers will work w/ you to design & construct labels to fit any size, permanent or removable adhesive, direct thermal or thermal transfer printable, can withstand extreme temperatures, harsh chemicals & abrasion. Labels can indicate sterilization such as autoclave or gamma or can be pre-sterilized. 100% synthetic, non-particulating, ultra low-outgassing raw materials wont contaminate clean or sterile environments. Lgis exclusive EZ-Peel feature is adhesive-free on one or both sides of the label, making it easy to apply & remove w/ protective gloves typical in cleanrooms, fabs, medical labs & other controlled environments.