Learn More
Uline Cleanroom Tape - 2"" x 36 yards, White, 24 rolls/case
Catalog No. NC1672656
Encompass
Seal bags and packages or mark floors and surfaces in biotech and medial labs. For Class 1 (ISO Class 3) cleanrooms. High-tack adhesive sticks firmly, removes cleanly. Polyethylene backing with rubber adhesive. Temperature range -20¿ to 200¿F.
Catalog No. NC1672656 Supplier Uline Supplier No. S20218WPK
Seal bags and packages or mark floors and surfaces in biotech and medial labs. For Class 1 (ISO Class 3) cleanrooms. High-tack adhesive sticks firmly, removes cleanly. Polyethylene backing with rubber adhesive. Temperature range -20¿ to 200¿F.