Learn More
CritiCore Protective Wear Cleanroom Boot
Catalog No. 18111280
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-111-280 Small
18-111-281 Medium
18-111-282 Large
18-111-283 X-Large
18-111-284 2X-Large
18-111-285 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 18-111-280 Supplier CritiCore Protective Wear Supplier No. 14M12141ESD01SWHT
This item is not returnable. View return policy
These reusable boots are suitable for a variety of cleanroom applications including: pharmaceutical, biotechnology, and medical device manufacturing.
  • High-density, ESD fabric for electrostatic management
  • Elastic at Ankle
  • Single Strap
  • Gamma sterilization stable gray molded sole
  • Color-coded strap for easy size identification
  • Laser-cut, pre-serged seams
  • Silicon-free
  • One reinforced snap on back of boot for coverall leg attachment
Specifications
Closure Type Single-strap Tunneled Elastic with Snaps
Color White
Material High Density ESD Polyester
Size Small
Sole Features Proprietary Molded Sole
Disposable Reusable
Sterility Non-sterile
Latex Free Yes