Learn More
CritiCore Protective Wear Cleanroom Boot
Description
- High-density, ESD fabric for electrostatic management
- Elastic at Ankle
- Single Strap
- Gamma sterilization stable gray molded sole
- Color-coded strap for easy size identification
- Laser-cut, pre-serged seams
- Silicon-free
- One reinforced snap on back of boot for coverall leg attachment
Specifications
Specifications
| Closure Type | Single-strap Tunneled Elastic with Snaps |
| Color | White |
| Material | High Density ESD Polyester |
| Size | Large |
| Sole Features | Proprietary Molded Sole |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Latex Free | Yes |