Description
- Cleanroom dry erase marker, bullet tip.
- Our Cleanroom Dry Erase Whiteboard markers are Class 10 certified and are made with an alcohol base for low odor.
- These dry erase markers work on whiteboards and most non-porous surfaces.
- They come in packs of 12 and are double bagged.
- The ink can be erased without solution using a dry wipe.
Specifications
Specifications
| Material | Plastic |
| Color | Black |
| Product Type | Cleanroom Dry Erase Marker |