Learn More
LGI EZ-Peel 2" x 1" Labels, 1000 pack
Catalog No. 501126973
Encompass_Preferred
This item has a minimum qty of 10 packs per supplier requirements.
leanMark 2" x 1" labels w/ deadened adhesive from Lgi. ISO 9001-2000 certified. RoHS compliant. 1k labels/roll. Contact your FisherSci rep for pricing/mins. Lgi also creates innovative custom labels that meet your specific cleanroom requirements. Based on their proprietary CleanMark process, engineers & designers will work w/ you to design & construct labels to fit any size, permanent or removable adhesive, direct thermal or thermal transfer printable, can withstand extreme temps, harsh chemicals & abrasion. Labels can indicate sterilization such as autoclave or gamma or can be pre-sterilized. Lgis exclusive EZ-Peel feature is adhesive-free on one or both sides of the label, making it easy to apply & remove w/ protective gloves typical in cleanrooms, fabs, medical labs & other controlled environments.
Catalog No. 50-112-6973 Supplier LGI Supplier No. 66011ORDER 10 OR MORE
This item has a minimum qty of 10 packs per supplier requirements.
leanMark 2" x 1" labels w/ deadened adhesive from Lgi. ISO 9001-2000 certified. RoHS compliant. 1k labels/roll. Contact your FisherSci rep for pricing/mins. Lgi also creates innovative custom labels that meet your specific cleanroom requirements. Based on their proprietary CleanMark process, engineers & designers will work w/ you to design & construct labels to fit any size, permanent or removable adhesive, direct thermal or thermal transfer printable, can withstand extreme temps, harsh chemicals & abrasion. Labels can indicate sterilization such as autoclave or gamma or can be pre-sterilized. Lgis exclusive EZ-Peel feature is adhesive-free on one or both sides of the label, making it easy to apply & remove w/ protective gloves typical in cleanrooms, fabs, medical labs & other controlled environments.