Learn More
LGI CLEANROOM EZ-PEEL ORNG LABEL
Catalog No. 501126970
Encompass_Preferred
This item has a minimum qty of 6 per supplier requirements.
CleanMark 2x1 labels from Lgi with deadened adhesive. ISO 9001-2000 certified. RoHS compliant. 1k labels per roll.Contact your FisherSci rep for pricing/mins. Lgi also creates innovative custom labels that meet your specific cleanroom requirements. Based on Lgis industry leading CleanMark, engineers & designers work with you to design & construct labels to fit any size, permanent or removable adhesive, direct thermal or thermal transfer printable, can withstand extreme temps, harsh chemicals & abrasion. Labels can indicate sterilization such as autoclave or gamma or can be pre-sterilized. 100% synthetic, non-particulating, ultra low-outgassing raw materials wont contaminate clean or sterile environments. Lgis exclusive EZ-Peel feature is adhesive-free on one or both sides of the label, making it easy to apply & remove with protective gloves typical in cleanrooms, fabs, medical labs & other controlled environments.
Catalog No. 50-112-6970 Supplier LGI Supplier No. 66014
This item has a minimum qty of 6 per supplier requirements.
CleanMark 2x1 labels from Lgi with deadened adhesive. ISO 9001-2000 certified. RoHS compliant. 1k labels per roll.Contact your FisherSci rep for pricing/mins. Lgi also creates innovative custom labels that meet your specific cleanroom requirements. Based on Lgis industry leading CleanMark, engineers & designers work with you to design & construct labels to fit any size, permanent or removable adhesive, direct thermal or thermal transfer printable, can withstand extreme temps, harsh chemicals & abrasion. Labels can indicate sterilization such as autoclave or gamma or can be pre-sterilized. 100% synthetic, non-particulating, ultra low-outgassing raw materials wont contaminate clean or sterile environments. Lgis exclusive EZ-Peel feature is adhesive-free on one or both sides of the label, making it easy to apply & remove with protective gloves typical in cleanrooms, fabs, medical labs & other controlled environments.