Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Cleanroom Pens

Catalog No. 18001511 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Blue
Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-001-511 Blue
18-001-507 Black
18-001-508 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-001-511
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed with low sodium ink to prevent contamination in cleanroom settings. Fisherbrand™ Cleanroom Pens come in black, blue, and red and feature HDPE construction.

  • Manufactured with very low sodium inks to prevent contamination
  • Barrel and cap made from high density polyethylene (HDPE)
  • Pens are packaged 10 per pack
  • Available in Black (18-001-507), Blue (18-001-511), or Red (18-001-508)

Specifications

Material HDPE
Color Blue
For Use With (Application) Cleanroom
Product Type Cleanroom Pen
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.