Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CritiCore Protective Wear Long Sleeve Cleanroom Scrub Top

Catalog No. 18111221
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-111-221 6X-Large
18-111-212 X-Small
18-111-213 Small
18-111-214 Medium
18-111-215 Large
18-111-216 X-Large
18-111-217 2X-Large
18-111-218 3X-Large
18-111-219 4X-Large
18-111-220 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 18-111-221 Supplier CritiCore Protective Wear Supplier No. 12TP2039SWSH01NVY6XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Long sleeve scrub tops are made from non-linting, breathable fabric and are ideal base-layers in controlled environments.

  • Non-linting
  • Continuous filament polyester
  • Pullover design
  • 3-Snap henley collar
  • Raglan sleeves
  • Side slits for added mobility
  • Filament knit cuffs

Specifications

Size 6X-Large
Material Sandwash Polyester
Color Navy
Neck Style Henley
Seam Type Pre-serged, Felled
Sleeve Style Raglan Sleeve
Closure Type 3 Snaps
Cuff Style Knit Cuff
Disposable Reusable
No. of Pockets None
Product Type Cleanroom Scrub Top
Sterility Non-sterile
For Use With (Application) Controlled Environments
Length (English) Sleeve 26 in.
Length (Metric) 66.04 cm
Width (English) 78 in.
Width (Metric) 198.12 cm
Height (English) 33 in.
Height (Metric) 83.82 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.