Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Cleanroom Sign Sleeve
SDP

Catalog No. 22000673
Change view
Click to view available options
Mounting Type:
Magnetic
Self-adhesive
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
22-000-673 Magnetic
22-000-674 Self-adhesive
22-000-675 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-000-673 Supplier Poltex Supplier No. CLEANSIGM
Add to Cart
Add to Cart
This item is not returnable. View return policy

Poltex sign sleeve is suitable for clean room applications.

  • PETG sign sleeve.
  • Suitable for clean room applications.
  • Front face lifts off for easy, thorough cleaning.
  • Choose from screw mount, magnet mount, or VHB Tape mount.

Sign sleeve, cleanroom, clean room, Poltex

Specifications

Material Polycarbonate
Size (English) 8.5 x 11 in.
Mounting Type Magnetic
For Use With (Application) Clean Room Applications
Shape Rectangular
Languages Universal
Product Type Sign Sleeve
Color Clear
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.