Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Cleanroom Sock Recycling Solution
GREENER_CHOICE

Catalog No. 16995939 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
12 x 11 x 38 in.
12 x 12 x 20 in.
15 x 13 x 38 in.
Capacity (Metric):
47.20 L
82.20 L
121.43 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Capacity (Metric)
16-995-939 12 x 12 x 20 in. 47.20 L
16-995-940 12 x 11 x 38 in. 82.20 L
16-995-941 15 x 13 x 38 in. 121.43 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16-995-939
Add to Cart
Add to Cart

By participating in the Fisherbrand Cleanroom Sock Recycling Solution program, your cleanroom socks will be diverted from the landfill. The box is made from recycled materials and includes a prepaid shipping label. Fill it at your own rate and ship it back for recycling when full.

  • Recycling solution for disposable cleanroom socks
  • All brands of disposable cleanroom socks are accepted
  • Eco-friendly Boxes: Made from recycled materials and easy to assemble
  • Collection boxes include a self-locking mechanisms and prepaid shipping label
  • Do not recycle biological materials, infectious waste, bloodborne pathogens, hazardous materials or socks exposed to more than 10% chemical exposure of the surface area
  • Available in 3 sizes: Small 12 x 12 x 20 in., Medium 12 x 11 x 38 in., Large 15 x 13 x 38 in.
  • 5 boxes per case
  • Quarterly report available upon request: Reports to include, but not limited to, materials recycled, amount processed, and materials end-of-life scenarios.

Specifications

Capacity (Metric) 47.20 L
Capacity (English) 10.38 gal.
Dimensions (L x W x H) 12 x 12 x 20 in.
Size Small
Autoclavable Not Autoclavable
For Use With (Equipment) Cleanroom Socks
Material Recycled Materials
Product Type Recycling Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.