Learn More
Med Vet International Waterless Surgical Scrub 3M Avagard 16oz Dispenser Refill Bottle 1% / 61% Strength Chg (Chlorhexidine Gluconate) / Ethyl Alcohol Nonsterile, 9200, 8/Case
Catalog No. 502829064
Encompass_Preferred
3M™ Avagard™ instant hand antiseptic with moisturizers is approved by the FDA The brushless and waterless refill works on contact, providing immediate microbial protection The alcohol quickly kills microorganisms to prevent infection and cross-contamination CHG, an antiseptic, offers cumulative activity with repeat use It moisturizes to prevent dryness and forms a protective barrier on the skin
Catalog No. 50-282-9064 Supplier Med Vet International Supplier No. 437989CS
3M™ Avagard™ instant hand antiseptic with moisturizers is approved by the FDA The brushless and waterless refill works on contact, providing immediate microbial protection The alcohol quickly kills microorganisms to prevent infection and cross-contamination CHG, an antiseptic, offers cumulative activity with repeat use It moisturizes to prevent dryness and forms a protective barrier on the skin