Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CleanSpace™ Technology Head Harness

Catalog No. 17800630 Shop All Cleanspace Technology Products
Change view
Click to view available options
For Use With (Equipment):
PAF-1034, PAF-2034
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17-800-630 PAF-1034, PAF-2034
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-800-630 Supplier CleanSpace™ Technology Supplier No. PAF0030
This item is not returnable. View return policy

Head harness is made of fabric and attached to the respirator unit using velcro strips.

  • Compatible with the CleanSpace2™ PAPR (PAF-0034)
  • Used with the revolutionary CleanSpace™ Respirator: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and offers breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement

Specifications

Certifications/Compliance AS/NZS1716: 2012, EN 12942 (CE Mark)
Disposable Reusable
Material Polyester, Nylon, Velcro
Product Type Head Harness
For Use With (Equipment) PAF-1034, PAF-2034
Model PAF-0030
Storage Requirements Store at -10°C to +55°C
Dimensions (L x W x H) 580 x 220 mm
Weight (Metric) 69 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.