Learn More
CleanSpace Technology™ CleanSpace3™ Cleaning and Storage Plug Set
Catalog No. 17800654 Shop All Cleanspace Technology Products
Change view
Click to view available options
For Use With (Equipment):
CleanSpace™ HALO Respirator unit (CS3000)
Catalog No. For Use With (Equipment)
17-800-654 CleanSpace™ HALO Respirator unit (CS3000)
1 options
This item is not returnable. View return policy
Do not wear the respirator when the plug set is in place.
Catalog No. 17-800-654 Supplier CleanSpace Technology™ Supplier No. CS3011
Add to Cart
Add to Cart
This item is not returnable. View return policy
Do not wear the respirator when the plug set is in place.
Tool for use with the CleanSpace™ HALO Respirator unit (CS3000). Seals all the openings on the CleanSpace™ HALO PAPR unit for storage. Made of polycarbonate and silicone.
  • Used with the revolutionary CleanSpace HALO PAPR: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
Specifications
Model CS3011
For Use With (Equipment) CleanSpace™ HALO Respirator unit (CS3000)