Learn More
CleanSpace™ HALO Full Face Mask for PAPR Assembly
Catalog No. 17800650 Shop All Cleanspace Technology Products
Change view
Click to view available options
Size:
Medium/Large
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-800-650 Small
17-800-651 Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-800-650 Supplier CleanSpace™ Supplier No. CS3006/DUP
Add to Cart
Add to Cart
Compatible with CleanSpace HALO Respirators, full face masks clip quickly onto the power units and provide ideal panoramic vision. Inner orinasal masks are made of soft silicone that readily molds to the face and does not cause allergic dermatitis.
  • Used with the revolutionary CleanSpace HALO: lightweight, no hoses or belts
  • Wide panoramic visor
  • Hard scratch and solvent resistant coating, age resistant materials
  • Suitable for long periods of wear
  • Built-in portacount adaptor for mask fit testing
  • Positive pressure airflow and inner orinasal mask prevents fogging
  • Easy breathing and no resistance even during heavy exertion
  • Visor seal silicone design seals to the face for optimum protection and comfort.
  • Inner Orinasal mask with unique clip-in clip-out design is easily removed for cleaning
  • Mylar speech diaphragm for clear communication on the job
Specifications
Certifications/Compliance AS/NZS1716: 2012
Certifications/Compliance (Europe) EN 12942
Product Type PAPR Full Face Mask
Size Small
Facepiece/Headtop Model Full face mask only
For Use With (Application) Healthcare, laboratory, pharmaceutical and emergency response
Intrinsic Safety (IS) Rating No
Model CS3006
If you require intrinsically safe equipment, please consult your site Intrinsically Safe approvals. These Half Masks are NOT certified intrinsically safe.