Learn More
CleanSpace™ HALO Half Mask for PAPR Assembly
Catalog No. 17800647 Shop All Cleanspace Technology Products
Change view
Click to view available options
Size:
Small
Catalog No. Size
17-800-647 Small
1 options
This item is not returnable. View return policy
Catalog No. 17-800-647 Supplier CleanSpace™ Supplier No. CS3003
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifically designed to work to complete the CleanSpace HALO system and clip quickly onto the power units. Half masks are made of soft silicone that readily molds to the face and does not cause allergic dermatitis.
  • Used with the revolutionary CleanSpace HALO: light weight, no hoses or belts
  • Half masks available in 3 sizes to facilitate a good seal and a comfortable fit
  • Materials: Soft silicone cushion, nylon clips and polycarbonate (exhalation valve)
  • Mask Fit Test adaptor available for TSI Portacount™ fit testing
  • Easy removal
  • Compatible with most cleaning and disinfection protocols
  • Transparent and facilitates communication
Specifications
Certifications/Compliance AS/NZS1716: 2012
Certifications/Compliance (Europe) EN 12942
Product Type PAPR Half Mask
Size Small
Facepiece/Headtop Model Half mask only
For Use With (Application) Primary healthcare facilities, pharmaceutical production, laboratories, animal research facilities and emergency/disaster responders
Intrinsic Safety (IS) Rating No
Model CS3003
If you require intrinsically safe equipment, please consult your site Intrinsically Safe approvals. These Half Masks are NOT certified intrinsically safe.