Learn More
CleanSpace™ Technology CleanSpace3™ Head Harness for Half Mask (non fabric)
Catalog No. 17800657 Shop All Cleanspace Technology Products
Change view
Click to view available options
For Use With (Equipment):
CS3000 PAPR System
Catalog No. For Use With (Equipment)
17-800-657 CS3000 PAPR System
1 options
Catalog No. 17-800-657 Supplier CleanSpace™ Technology Supplier No. CS3008
Add to Cart
Add to Cart
Replacement part for use with the CleanSpace HALO. Consists of TPU clips, polyethylene strap with polyester adjustment band.
  • Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Multiple adjustment points
  • Easy to wash and quick drying
  • Durable and designed for long wear in harsh environments
  • Easy and quick replacement
Specifications
Disposable Reusable
Product Type Head Harness
For Use With (Equipment) CS3000 PAPR System
Model CS3008