Learn More
CleanSpace™ Technology CleanSpace™ HALO Neck Support
Catalog No. 17800659 Shop All Cleanspace Technology Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-800-659 Large
17-800-658 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-800-659 Supplier CleanSpace™ Technology Supplier No. CS3010
Add to Cart
Add to Cart
Plastic neck support is for use with the CleanSpace HALO PAPR units.
  • Used with CleanSpace respirators: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
Specifications
Disposable Reusable
Material ABS Plastic
Product Type Neck Support
Size Large
For Use With (Equipment) CS3000 PAPR System
Model CS3010