Learn More
MSC CleanTeam Cleanroom Latex Glove, Class 100, Textured Grip, 12 in
Catalog No. 5000792084
Encompass
These single use gloves are designed for controlled environments requiring Class 100 cleanroom compatibility. Made from latex, they feature a fully textured grip and a rolled cuff for secure wear and enhanced dexterity.
  • ISO 5 (Class 100) cleanroom compatibility
  • Latex polymer construction
  • Fully textured grip on palm and fingers
  • Rolled cuff for added strength and easier donning
  • Extended 12 in length
  • Powder-free design
  • Optimal 7 mil thickness
  • Available in sizes S-2XL
  • Packed with 10 bags per case
Catalog No. 50-007-92084 Supplier MSC Supplier No. 76000793
These single use gloves are designed for controlled environments requiring Class 100 cleanroom compatibility. Made from latex, they feature a fully textured grip and a rolled cuff for secure wear and enhanced dexterity.
  • ISO 5 (Class 100) cleanroom compatibility
  • Latex polymer construction
  • Fully textured grip on palm and fingers
  • Rolled cuff for added strength and easier donning
  • Extended 12 in length
  • Powder-free design
  • Optimal 7 mil thickness
  • Available in sizes S-2XL
  • Packed with 10 bags per case