Learn More
Electron Microscopy Sciences Cleanware Aqua-Clear Water Conditioner 100 ML
SDP
Catalog No. 50997490
Encompass_Preferred
Aqua-Clear Water Conditioner stabilizes water in water baths and other water systems. Just 2 ml per liter of water provides long lasting protection against the growth of algae, bacteria, and molds which often contaminate standing water. It also protects pumps and thermostats and helps prevent tubing from clogging. In addition, it may be added to the water that is used in developers for color or X-ray films. It will not harm the film and permits longer use of the same solution.
Aqua-Clear Water Conditioner adds a blue tint to the water which will fade after about 3 months reminding you to change the water and add new conditioner.
Catalog No. 50-997-490 Supplier Electron Microscopy Sciences Supplier No. 6050201
Aqua-Clear Water Conditioner stabilizes water in water baths and other water systems. Just 2 ml per liter of water provides long lasting protection against the growth of algae, bacteria, and molds which often contaminate standing water. It also protects pumps and thermostats and helps prevent tubing from clogging. In addition, it may be added to the water that is used in developers for color or X-ray films. It will not harm the film and permits longer use of the same solution.
Aqua-Clear Water Conditioner adds a blue tint to the water which will fade after about 3 months reminding you to change the water and add new conditioner.