Learn More
Analytical Sales & Services Clear 100µL Low Profile PCR Plate - No Skirt
Catalog No. 5000805815
Encompass
Advantage Clear 100µL Low Profile PCR Plate No Skirt. Thin Walled - Easy to Cut Into Sections.\n\nQty/Pkg: 100
Catalog No. 50-008-05815 Supplier Analytical Sales & Services Supplier No. 969619
Advantage Clear 100µL Low Profile PCR Plate No Skirt. Thin Walled - Easy to Cut Into Sections.\n\nQty/Pkg: 100