Learn More
Analytical Sales & Services Clear 200µL PCR Plate - Flat Well Rim, No Skirt
Catalog No. 5000806107
Encompass
Clear 200µL PCR Plate. Flat Well Rim Thin Walled Can Be Cut Into Sections\n\nQty/Pkg: 100
Catalog No. 50-008-06107 Supplier Analytical Sales & Services Supplier No. 969610
Add to Cart
Add to Cart
Clear 200µL PCR Plate. Flat Well Rim Thin Walled Can Be Cut Into Sections\n\nQty/Pkg: 100