Learn More
Analytical Sales & Services Clear 200µL Raised Rim PCR Plate
Catalog No. 5000806197
Encompass
Clear 200μL PCR Plate, Wide skirt for Barcoding, 1mm Raised Rim Around Each Well.\n\nAlternative to Perkin-Elmer Plate.\n\nQty/Pkg: 100
Catalog No. 50-008-06197 Supplier Analytical Sales & Services Supplier No. 969601
Clear 200μL PCR Plate, Wide skirt for Barcoding, 1mm Raised Rim Around Each Well.\n\nAlternative to Perkin-Elmer Plate.\n\nQty/Pkg: 100