Learn More
Industrial Glassware CLEAR 40ML VIALS 288/CS
Catalog No. NC0409908
Encompass_Preferred
Pre-cleaned 40-ml clear glass VOA vial with 24-414 open-top septum cap. Barcoded label on vial referencing Lot Number. 288 vials per case (4 x 72).
Catalog No. NC0409908 Supplier Industrial Glassware Supplier No. 2795FLPC4
Pre-cleaned 40-ml clear glass VOA vial with 24-414 open-top septum cap. Barcoded label on vial referencing Lot Number. 288 vials per case (4 x 72).