Learn More
Greiner Bio-One μClear™ 96-well, Cell Repellent-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Catalog No. 07000640 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-640 Clear
1 options
Catalog No. 07-000-640 Supplier Greiner Bio-One Supplier No. 655976
Add to Cart
Add to Cart
Color:Clear; Sterility:Sterile; Lid:With Lid
  • Characterized by low cell attachment
  • ideal for applications such as suspension culture of semi-adherent and adherent cells, Gel-based 3D cultures and Magnetic 3D cell culturing (Nano3D technology)
  • μClear coated well bottom
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid With Lid
Array 8 x 12
Packaging 8/Pk., 32/Cs.
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Cell Repellent
Well Shape Flat Bottom (F-well)
No. per Case 32
Quantity 32/Cs.