Learn More
Alkali Scientific Western Blot Boxes, 3 3/4 X 1 3/16 X 5/8", Min Volume 2mL, Clear, 5/pk
SDP
Catalog No. NC1142595
Encompass_Preferred
Western blot boxes and containers are available in clear. They fit nearly any sized blot and feature a hinged lid that easy to open and close. The corners are 90˚ and they have a flatter and more uniform surface. Use less reagents and antibodies with these cost-efficient western blot boxes. When you match the size of the box with your blot you're able to fit more blots on your rocker and shaker allowing for a smaller blot footprint.
Catalog No. NC1142595 Supplier Alkali Scientific Supplier No. BC12009
Western blot boxes and containers are available in clear. They fit nearly any sized blot and feature a hinged lid that easy to open and close. The corners are 90˚ and they have a flatter and more uniform surface. Use less reagents and antibodies with these cost-efficient western blot boxes. When you match the size of the box with your blot you're able to fit more blots on your rocker and shaker allowing for a smaller blot footprint.