Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Boston Round Bottles with Black Phenolic Rubber Caps

Catalog No. 01001478
Change view
Click to view available options
Capacity (Metric):
950 mL
Quantity:
30/Cs., 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
01-001-478 950 mL 30/Cs., 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-478 Supplier Qorpak™ Supplier No. 276161

missing translation for 'validMainframeMsg'

General-use bottles for liquids, product storage, and field or plant sampling projects. Qorpak™ Clear Boston Round Bottles with Black Phenolic Rubber Cap offer maximum visibility and sample integrity.

  • Features a round body, curved shoulders and narrow screw neck opening
  • Environmental sensitive help eliminate waste and ensure product integrity for long term storage
  • Glass offers minimal chemical interference with contents

Specifications

Capacity (Metric) 950 mL
CAS 75-05-8, 17834-03-6
Color Clear
Material Glass (Type 3)
Linear Formula C19H16O5
MDL Number MFCD00272668
Concentration 1 mg/mL in acetonitrile
No. per Case 30/Cs.
Quantity 30/Cs., 1 mL
Sterility Non-sterile
Formula Weight 324.33
Closure Color Black
Packaging Ampule
Closure Included Yes
Graduated No
Closure Liner Rubber Liner
Closure Material Phenolic
Closure Size 33-400
Closure Type Continuous Thread
Capacity (English) 32 oz.
Shape Round
Disposable Reusable
Flash Point 6°C (42.8°F) - closed cup
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.