Learn More
Corning™ Clear Bottom Black Polystyrene TC-treated Microplates, 96 Half Area Well
GREENER_CHOICE
Catalog No. 01213107 Shop All Corning Products
Change view
Click to view available options
Color:
Clear, Black
Catalog No. Color
01-213-107 Clear, Black
1 options
This item is not returnable. View return policy
Catalog No. 01-213-107 Supplier Corning™ Supplier No. 3887A
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm
  • Opaque walls prevent well-to-well cross-talk
  • Optically clear flat bottom permits direct microscopic viewing
TRUSTED_SUSTAINABILITY
Specifications
Format Half-Area
Lid Without Lid
Culture Area 0.16 cm2
Array 12 x 8
Color Clear, Black
Volume (Metric) Well 50 to 100 μL
No. of Wells 96
Volume (Metric) Working 25 to 125 μL
Sterility Sterile
Surface Treatment Tissue Culture
Well Shape Flat Bottom (F-well)
No. per Case 100
No. per Pack 25
Well Color Clear
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov