Learn More
STA Pharmaceutical Masterflex Transfer Tubing, C-Flex, Clear, 1/4"" ID x 1/2"" OD; 25 ft
Catalog No. NC9373183
Encompass
Applications: Pharmaceutical, biotech, and general-purpose lab uses requiring high performance. Economical alternative to platinum silicone. Characteristics: Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and a smooth surface. Transparent. Certification: Product is manufactured in accordance with Part 820 Quality System Requirements (GMP); and meets USP Class VI. Temperature range: -40 to 275¿F (-40 to 135¿C) Sterilize: Autoclave, ethylene oxide, or gamma irradiation.
Catalog No. NC9373183 Supplier STA Pharmaceutical Supplier No. MFLX0642212
Applications: Pharmaceutical, biotech, and general-purpose lab uses requiring high performance. Economical alternative to platinum silicone. Characteristics: Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and a smooth surface. Transparent. Certification: Product is manufactured in accordance with Part 820 Quality System Requirements (GMP); and meets USP Class VI. Temperature range: -40 to 275¿F (-40 to 135¿C) Sterilize: Autoclave, ethylene oxide, or gamma irradiation.