Learn More
Electron Microscopy Sciences Clear Dropping Bottle, 50 ml
SDP
Catalog No. 5029848
Encompass_Preferred
These bottles deliver stains and indicator solutions drop by drop from a projecting drip tip on the side of the stopper. A partial turn of the ground glass stopper will seal the bottle.
Diameter: 45 mm
Height: 95 mm
Catalog No. 50-298-48 Supplier Electron Microscopy Sciences Supplier No. 6422005
These bottles deliver stains and indicator solutions drop by drop from a projecting drip tip on the side of the stopper. A partial turn of the ground glass stopper will seal the bottle.
Diameter: 45 mm
Height: 95 mm