Learn More
Alkali Scientific ASI™ Clear, Easy-Grip, 35 x10mm, Confocal Dish, Adhesion Type, Sterile, Non-Treated (Non-Charged/ Hydrophobic), 100/Cs, 22 x 22 x 0.13mm, Hole 3.14mm, PS/Glass
SDP
Catalog No. 503028859
Encompass_Preferred
ASI™ confocal products allow researchers to acquire high resolution microscopic images of cells in 35 mm culture dishes or 6 well plates. Low auto-fluorescence glass and FLux were used, ideal for uses of these products are confocal microscopy, phase contrast microscopy, live cell imaging and micro-manipulations. Surface materials for confocal products:Glass: naturally charged, good growth surface for adherent cellsFLux: surface modified plastic film (188 μm thick), optimal for confocal microscopy Bottom materials: Glass/FLux (no surface treatment for glass bottom) Coverslip: #1.5 Dish color: Clear/Black Dish size: 35 Ø Hole size: 13 Ø / 20 Ø / 25Ø Non – pyrogenic Non – cytotoxic DNase / RNase – free Human DNA – free  
Catalog No. 50-302-8859 Supplier Alkali Scientific Supplier No. TCG23
ASI™ confocal products allow researchers to acquire high resolution microscopic images of cells in 35 mm culture dishes or 6 well plates. Low auto-fluorescence glass and FLux were used, ideal for uses of these products are confocal microscopy, phase contrast microscopy, live cell imaging and micro-manipulations. Surface materials for confocal products:Glass: naturally charged, good growth surface for adherent cellsFLux: surface modified plastic film (188 μm thick), optimal for confocal microscopy Bottom materials: Glass/FLux (no surface treatment for glass bottom) Coverslip: #1.5 Dish color: Clear/Black Dish size: 35 Ø Hole size: 13 Ø / 20 Ø / 25Ø Non – pyrogenic Non – cytotoxic DNase / RNase – free Human DNA – free  