Learn More
WELCH MATERIALS INC DoprahTM Clear Glass EPA/TOC Vial with Cap & Septa with Dust Cover(EPA Quality), pre-assembled, 27.5×95mm, 72pcs/pk
Catalog No. 502093127
Encompass_Preferred
40mL 27.5x95mm Clear Glass EPA/TOC Vial 24-400 White Open TopPP Screw Cap with 22mm Natural PTFE/White Silicone 3.0mm thickSepta (EPA Quality) and Dust Cover, Pre-assembled. 72 pcs/pk.
Catalog No. 50-209-3127 Supplier WELCH MATERIALS INC Supplier No. DK1342
40mL 27.5x95mm Clear Glass EPA/TOC Vial 24-400 White Open TopPP Screw Cap with 22mm Natural PTFE/White Silicone 3.0mm thickSepta (EPA Quality) and Dust Cover, Pre-assembled. 72 pcs/pk.