Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
GSA_VA

Catalog No. 12565376
Change view
Click to view available options
Surface Treatment:
Enhanced Binding
Immulon 1 B
Immulon 2 HB
Immulon 4 HBX
MaxiSorp
MediSorp
MultiSorp
PolySorp
Universal Binding
Certified:
No
Yes
Volume (Metric) Working:
230 μL
280 μL
330 μL
350 μL
Barcode:
No Barcode
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment Certified Volume (Metric) Working Barcode
12-565-376 PolySorp No 350 μL No Barcode
14-245-78 Immulon 1 B No 230 μL No Barcode
14-245-61 Immulon 2 HB No 280 μL No Barcode
14-245-153 Immulon 4 HBX No 280 μL No Barcode
12-565-135 MaxiSorp Yes 350 μL No Barcode
12-565-136 MaxiSorp No 350 μL No Barcode
12-565-377 MaxiSorp Yes 350 μL No Barcode
12-565-730 MediSorp Yes 350 μL No Barcode
14-962-23 MultiSorp No 350 μL No Barcode
12-565-138 PolySorp No 350 μL No Barcode
21-377-269 Universal Binding No 330 μL No Barcode
21-377-270 Enhanced Binding Yes 330 μL No Barcode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Each lot is functionally tested for IgG binding.
Catalog No. 12-565-376 Supplier Thermo Scientific™ Supplier No. 456529
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Each lot is functionally tested for IgG binding.

Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.

Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

96-Well Solid Plates:

  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces:

  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.

Ordering Information:

Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certified No
Format Standard
Lid Without Lid
Detection Method Colorimetric
Array 8 x 12
Packaging 180 Cases
Binding Type Adsorption
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 350 μL
Sterility Non-sterile
Surface Treatment PolySorp
Well Shape Flat Bottom (F-well)
No. per Case 180
No. per Pack 10
Plate Blocking None
Quantity 180/Cs.
Target Molecule Lipids
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.