Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
GREENER_CHOICE

Catalog No. 21377270 Shop All Thermo Scientific Products
Change view
Click to view available options
Surface Treatment:
Enhanced Binding
Immulon 1 B
Immulon 2 HB
Immulon 4 HBX
MaxiSorp
MediSorp
MultiSorp
PolySorp
Universal Binding
Certified:
No
Yes
Volume (Metric) Working:
230 μL
280 μL
330 μL
350 μL
Barcode:
No Barcode
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment Certified Volume (Metric) Working Barcode
21-377-270 Enhanced Binding Yes 330 μL No Barcode
14-245-78 Immulon 1 B No 230 μL No Barcode
14-245-61 Immulon 2 HB No 280 μL No Barcode
14-245-153 Immulon 4 HBX No 280 μL No Barcode
12-565-135 MaxiSorp Yes 350 μL No Barcode
12-565-136 MaxiSorp No 350 μL No Barcode
12-565-376 PolySorp No 350 μL No Barcode
12-565-377 MaxiSorp Yes 350 μL No Barcode
12-565-730 MediSorp Yes 350 μL No Barcode
14-962-23 MultiSorp No 350 μL No Barcode
12-565-138 PolySorp No 350 μL No Barcode
21-377-269 Universal Binding No 330 μL No Barcode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Each lot is functionally tested for IgG binding.
Catalog No. 21-377-270 Supplier Thermo Scientific™ Supplier No. 95029330
  • $518.00
    $430.65 / Case of 50

    Save $87.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Each lot is functionally tested for IgG binding.

Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.

Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

96-Well Solid Plates:

  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces:

  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.

Ordering Information:

Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Hydrophilic
Certified Yes
Format Standard
Detection Method Colorimetric
Array 8 x 12
Packaging 50 Cases
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 450 μL
No. of Wells 96
Volume (Metric) Working 330 μL
Sterility Non-sterile
Surface Treatment Enhanced Binding
Well Shape Flat Bottom (F-well)
No. per Case 50
No. per Pack 25
Plate Blocking None
Target Molecule Antibodies (Glycoproteins)
Show More Show Less
For Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, is the surface treatment solely on the bottom of the well, or is it also applied to the sides?

For energy treated plates, the entire plate surface receives the energy treatment. The MaxiSorp, PolySorp, MediSorp and MultiSorp plates are energy treated plates. For traditionally treated plates, the well surface is treated up to the listed working volume.

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

Which Nunc immunodiagnostic passive surface should I choose: PolySorp, MediSorp, MaxiSorp, or MultiSorp?

Our passive immunodiagnostic surfaces are based on a series of treatments that modify the polystyrene surface to different final levels of hydrophilicity. Selection of one of these surfaces for ELISA development should be based on the characteristics of the biomolecule you seek to immobilize. PolySorp is fairly hydrophobic and should be used for biomolecules that have hydrophobic domains, e.g., lipids, lipoproteins, and large proteins. MediSorp is slightly hydrophilic and can be used for biomolecules with hydrophilic and hydrophobic properties, e.g., medium-to-large proteins such as albumin, or amphiphilic biomolecules such as LPS. MaxiSorp is quite hydrophilic, optimized for binding of IgG (and any other molecule with similar characteristics) and thus is a popular choice for sandwich ELISA and similar protocols. MultiSorp is very hydrophilic, and in turn will immobilize very hydrophilic biomolecules such as glycoproteins. Some optimization will always be necessary, and in many cases screening for binding optimization may involve a few surface types.

What is the difference between a certified and an uncertified immunodiagnostic plate, e.g., Cat. No. 439454 vs. 442404?

There is no difference in the manufacturing process. Certified plates are performance tested (using an antibody binding protocol) during quality control, whereas uncertified plates are not. This test is expensive for us to perform, so we offer the uncertified version at cost savings customers for whom antibody binding testing results are not critical.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.