Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Clear Fluid Pressure Apparatus
GREENER_CHOICESCI_ED

Catalog No. S139431 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
9+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139431 9+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139431 Supplier Supertek™ Supplier No. PH10627SP
Add to Cart
Add to Cart

The Clear Fluid Pressure apparatus demonstrates liquid pressure increasing with depth.

  • Includes:
  • 1 - Clear cylinder made from transparent thermo plastic.
  • Dimensions 15.75 x 2.36 in. (H x Dia.)
  • Instructions included

Specifications

Grade 9+
Format Model
Languages English
Subject Physical Science - Pressure and Volume
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.