Learn More
Waters Corp Clear Glass 15 x 75 mm Screw Neck Total Recovery Vial, 100/pk
Catalog No. 50727115 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 75 mm Total Recovery Vial is designed for use in the PATROL UPLC Process Analyzer and PATROL UPLC Laboratory Analyzer, where sample volumes are limited.
This item is not returnable. View return policy
Catalog No. 50-727-115 Supplier Waters Corp Supplier No. 186007573
The 75 mm Total Recovery Vial is designed for use in the PATROL UPLC Process Analyzer and PATROL UPLC Laboratory Analyzer, where sample volumes are limited.