Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Clear Glass Narrow Mouth Reagent Bottles with Glass Stoppers
GREENER_CHOICESCI_ED

Catalog No. S139266 Shop All Supertek Scientific Products
Change view
Click to view available options
Capacity (Metric):
50 mL
60 mL
100 mL
250 mL
500 mL
1000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S139266 60 mL
S139265 50 mL
S139267 100 mL
S139268 250 mL
S139269 500 mL
S139270 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. S139266 Supplier Supertek™ Supplier No. 12.101.0060SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

These reusable Clear Glass Narrow Mouth Reagent Bottles with Glass Stoppers are strong, chemical resistant bottles with an enameled area for marking.

  • In conformity to DIN ISO 4796-2 Standard
  • Printed in white color
  • Sturdy ground neck
  • All sizes have a enameled area for marking
  • Strong and chemical resistant
  • Supplied with solid glass stopper
  • Autoclavable

Specifications

Capacity 60 mL
Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Transparent
Diameter (English) 1.7 ±0.1 in.
For Use With (Application) Storage
Material Borosilicate Glass (3.3)
Quantity 12 Pk.
Sterility Sterile
Certifications/Compliance DIN ISO 4796-2
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 9 cm
Product Type Reagent Bottle
For Use With (Equipment) Chemicals
Height (English) 3.5 in.
Chemical Label Custom Write-on
Closure Liner Unlined
Closure Material Borosilicate Glass
Closure Size 14/23
Diameter (Metric) 42 ±2 mm
Closure Type Stopper
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.