Learn More
Supertek™ Clear Glass Narrow Mouth Reagent Bottles with Glass Stoppers

Shop All Supertek Scientific Products


Description
- In conformity to DIN ISO 4796-2 Standard
- Printed in white color
- Sturdy ground neck
- All sizes have a enameled area for marking
- Strong and chemical resistant
- Supplied with solid glass stopper
- Autoclavable
Specifications
Specifications
| Capacity | 250 mL |
| Capacity (Metric) | 250 mL |
| Capacity (English) | 8.5 oz. |
| Color | Transparent |
| Diameter (English) | 2.8 ±0.1 in. |
| For Use With (Application) | Storage |
| Material | Borosilicate Glass (3.3) |
| Quantity | 12 Pk. |
| Sterility | Sterile |
| Certifications/Compliance | DIN ISO 4796-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.