Learn More
Quality Lab Accessories 1000 mL Clear Glass Vessel, Borosilicate Glass, Dishwasher Safe, Each
Catalog No. NC3452498
Encompass
The 1000 mL Clear Glass Vessel with Plastic Rim is designed for durability in laboratory applications. Its borosilicate glass construction and high-impact plastic rim minimize breakage, ensuring reliable performance during dissolution testing. This vessel is dishwasher safe and comes with a Certificate of Compliance, making it a trusted choice for labs looking to maintain high standards of cleanliness and safety.
Catalog No. NC3452498 Supplier Quality Lab Accessories Supplier No. GLA900UNP
The 1000 mL Clear Glass Vessel with Plastic Rim is designed for durability in laboratory applications. Its borosilicate glass construction and high-impact plastic rim minimize breakage, ensuring reliable performance during dissolution testing. This vessel is dishwasher safe and comes with a Certificate of Compliance, making it a trusted choice for labs looking to maintain high standards of cleanliness and safety.