Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIMBLE™ Clear Glass Serum Vial Without Closure
SDP

Catalog No. 01672737 Shop All DWK Life Sciences Products
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
01-672-737 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-737 Supplier KIMBLE™ Supplier No. 62113D312
Add to Cart
Add to Cart

Serum vial with special blowback design for ease of use with lyophillization stoppers.

  • Converted from Type I borosilicate glass tubing
  • Serum vials are lighter in weight than molded bottles, with more closely held tolerances and uniformity of glass
  • Special blowback design provides ease of use for lyophillization stoppers
  • Aluminum seals, stoppers and septa are available separately
  • Special design provides extra strength for freeze-drying applications
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Diameter (Metric) Outer 18 mm
Material Borosilicate Glass (Type 1 Class A)
Pre-slit No
Sterility Non-sterile
Volume (Metric) 3.5 mL
Closure Type Not Included
Quantity 1440/Pk.
Marking Spot No
Autoclavable Autoclavable
Height (Metric) 45 mm
Product Type Serum Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.